Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ufm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026435.5 | NP_080711.1 | 6 | 272 | ||
GenBank mRNA | AK007788.1 | - | 6 | 236 | ||
AK008280.1 | - | 6 | 235 | |||
AK011737.1 | - | 6 | 252 | |||
AK089948.1 | - | 6 | 238 | |||
AK148146.1 | - | 6 | 242 | |||
AK166332.1 | - | 6 | 266 | |||
AK167301.1 | - | 6 | 241 | |||
AK167321.1 | - | 6 | 236 | |||
BC051554.1 | - | 5 | 236 | |||
BC061065.1 | - | 6 | 271 | 6594260 | ||
CJ075089.1 | - | 6 | 298 |