Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtfp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026443.4 | NP_080719.2 | 4 | 1369 | ||
GenBank mRNA | AK010971.1 | - | 4 | 1326 | ||
AK012058.1 | - | 4 | 1336 | |||
AK078381.1 | - | 4 | 1370 | |||
AK167990.1 | - | 4 | 1238 | |||
AK216258.1 | - | 1 | 339 | |||
BC012242.1 | - | 1 | 783 | |||
BC019205.1 | - | 4 | 1379 | |||
BC058737.1 | - | 4 | 1319 | 5400435 |