Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1600012H06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083880.1 | NP_001077349.1 | 2 | 430 | ||
NM_001083881.1 | NP_001077350.1 | 2 | 500 | |||
NM_001083882.1 | NP_001077351.1 | 1 | 682 | |||
NM_026451.2 | NP_080727.2 | 2 | 435 | |||
GenBank mRNA | AK005427.1 | - | 2 | 412 | ||
AK017276.1 | - | 1 | 612 | |||
AK018298.1 | - | 2 | 358 | |||
AK028052.1 | - | 2 | 436 | |||
AK076083.1 | - | 2 | 392 | |||
AK076441.1 | - | 639 | ||||
AK076476.1 | - | 2 | 355 | |||
AK083064.1 | - | 1 | 623 | |||
AK150772.1 | - | 2 | 455 | |||
AK167490.1 | - | 2 | 442 | |||
BB610149.1 | - | 2 | 436 | |||
BC027313.1 | - | 2 | 414 | 3710655 | ||
BC043303.1 | - | 1 | 643 | |||
BC043449.1 | - | 1 | 643 | |||
BC054359.1 | - | 2 | 432 | |||
BC071250.1 | - | 2 | 347 |