Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prorsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163454.2 | NP_001156926.1 | 3 | 747 | ||
NM_026465.3 | NP_080741.1 | 2 | 656 | |||
GenBank mRNA | AK008574.1 | - | 2 | 657 | ||
AK077280.1 | - | 3 | 746 | |||
BC033930.1 | - | 3 | 720 | 1397170 |