Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps27l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026467.3 | NP_080743.1 | 4 | 360 | ||
GenBank mRNA | AK007646.1 | - | 1 | 67 | ||
AK007689.1 | - | 4 | 361 | |||
AK011262.1 | - | 4 | 379 | |||
AK151083.1 | - | 4 | 379 | |||
BC058115.1 | - | 4 | 361 | 6407738 |