Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tppp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026481.3 | NP_080757.1 | 3, 4 | 498 | ||
GenBank mRNA | AK009771.1 | - | 3, 4 | 462 | ||
AK012437.1 | - | 3, 4 | 466 | |||
AK155018.1 | - | 3, 4 | 469 | |||
AK160507.1 | - | 3, 4 | 469 | |||
BC010788.1 | - | 3, 4 | 455 | 4216285 | ||
BC116622.1 | - | 2, 3 | 290 |