Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccny | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026484.3 | NP_080760.2 | 1, 2 | 625 | ||
GenBank mRNA | AK014201.1 | - | 1, 2 | 630 | ||
AK017493.1 | - | 1, 2 | 457 | |||
AK028696.1 | - | 1, 2 | 626 | |||
AK076556.1 | - | 1, 2 | 155 | |||
AK178229.1 | - | 1, 2 | 64 | |||
BC023321.1 | - | 2, 3 | 543 | 3995040 |