Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trabd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026485.2 | NP_080761.2 | 7, 8 | 781 | ||
GenBank mRNA | AK005300.1 | - | 7, 8 | 751 | ||
AK017745.1 | - | 7, 8 | 775 | |||
AK146027.1 | - | 7, 8 | 797 | |||
AK185880.1 | - | 341 | ||||
BC005574.1 | - | 7, 8 | 807 | 3967961 | ||
BU510197.1 | - | 7, 8 | 861 |