Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Murc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026509.3 | NP_080785.2 | 1 | 460 | ||
GenBank mRNA | AK009691.1 | - | 1 | 417 | ||
BC089601.1 | - | 1 | 460 | 6788090 | ||
EU487254.1 | - | 1 | 352 |