Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam122a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026520.3 | NP_080796.1 | 1 | 1310 | ||
GenBank mRNA | AK018219.1 | - | 1 | 1261 | ||
AK033646.1 | - | 1 | 1310 | |||
AK077654.1 | - | 1 | 1293 | |||
AK172089.1 | - | 1 | 1295 | |||
BC028637.1 | - | 1 | 1278 | 1248203 |