Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp706 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026521.4 | NP_080797.1 | 4 | 538 | ||
GenBank mRNA | AB041652.1 | - | 4 | 481 | ||
AK004076.1 | - | 4 | 337 | |||
AK032231.1 | - | 4 | 424 | |||
AK082392.1 | - | 4 | 442 | |||
AK132053.1 | - | 4 | 455 | |||
AK145319.1 | - | 4 | 416 | |||
AK166530.1 | - | 4 | 416 | |||
AK167791.1 | - | 4 | 526 | |||
AK167897.1 | - | 4 | 455 | |||
AK168303.1 | - | 4 | 455 | |||
AK168958.1 | - | 4 | 417 | |||
BC037600.1 | - | 4 | 463 | |||
BC042704.1 | - | 4 | 429 | |||
BC052459.1 | - | 4 | 538 | |||
BC054356.1 | - | 4 | 460 | |||
BC085477.1 | - | 4 | 422 | |||
BC147306.1 | - | 4 | 397 | 9056379 | ||
BC147307.1 | - | 4 | 397 | 9056367 |