Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp706 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026521.4 | NP_080797.1 | 4 | 949 | ||
GenBank mRNA | AB041652.1 | - | 4 | 892 | ||
AK032231.1 | - | 4 | 835 | |||
AK082392.1 | - | 4 | 853 | |||
AK132053.1 | - | 4 | 866 | |||
AK145319.1 | - | 4 | 827 | |||
AK166530.1 | - | 4 | 827 | |||
AK168958.1 | - | 4 | 828 | |||
AK215792.1 | - | 1 | 127 | |||
BC052459.1 | - | 4 | 949 | |||
BC085477.1 | - | 4 | 833 | |||
BC147306.1 | - | 4 | 808 | 9056379 | ||
BC147307.1 | - | 4 | 808 | 9056367 |