Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp706 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026521.4 | NP_080797.1 | 4 | 1476 | ||
GenBank mRNA | AB041652.1 | - | 4 | 1419 | ||
AK014015.1 | - | 1 | 817 | |||
AK032231.1 | - | 4 | 1362 | |||
AK082392.1 | - | 4 | 1380 | |||
AK132053.1 | - | 4 | 1393 | |||
AK145319.1 | - | 4 | 1354 | |||
AK166530.1 | - | 4 | 1354 | |||
AK167897.1 | - | 4 | 1393 | |||
AK168958.1 | - | 4 | 1355 | |||
AK200355.1 | - | 1 | 363 | |||
BC037600.1 | - | 4 | 1401 | |||
BC042704.1 | - | 4 | 1367 | |||
BC052459.1 | - | 4 | 1476 | |||
BC054356.1 | - | 4 | 1398 | |||
BC085477.1 | - | 4 | 1360 | |||
BC147306.1 | - | 4 | 1335 | 9056379 | ||
BC147307.1 | - | 4 | 1335 | 9056367 |