Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142681.1 | NP_001136153.1 | 13 | 1290 | ||
NM_026522.5 | NP_080798.2 | 11 | 1032 | |||
GenBank mRNA | AK014071.1 | - | 13 | 1239 | ||
AK036717.1 | - | 13 | 1235 | |||
AK050655.1 | - | 11 | 980 | |||
AK147377.1 | - | 11 | 6088 | |||
AK160156.1 | - | 13 | 1250 | |||
AK160200.1 | - | 13 | 1251 | |||
BC006876.1 | - | 13 | 1239 | 2647540 | ||
BC061063.1 | - | 13 | 1250 | 6591338 |