Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: N6amt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026526.2 | NP_080802.1 | 5 | 694 | ||
GenBank mRNA | AK005312.1 | - | 5 | 688 | ||
AK010911.1 | - | 4 | 689 | |||
AK012083.1 | - | 5 | 695 | |||
BC051925.1 | - | 5 | 685 | 5691329 |