Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290667.1 | NP_001277596.1 | 2 | 990 | ||
NM_026527.3 | NP_080803.1 | 3 | 1026 | |||
GenBank mRNA | AK007629.1 | - | 3 | 950 | ||
AK010914.1 | - | 3 | 996 | |||
AK021391.1 | - | 2 | 960 | |||
AK078993.1 | - | 3 | 1012 | |||
BC025100.1 | - | 3 | 940 | 3967723 |