Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026533.3 | NP_080809.1 | 5 | 460 | ||
GenBank mRNA | AK012479.1 | - | 5 | 405 | ||
AK018703.1 | - | 5 | 404 | |||
AK076060.1 | - | 5 | 404 | |||
AK088237.1 | - | 4 | 1037 | |||
AK151043.1 | - | 5 | 405 | |||
AK151671.1 | - | 5 | 406 | |||
AK161798.1 | - | 3 | 1452 | |||
AK166691.1 | - | 5 | 405 | |||
AV498110.1 | - | 4 | 367 | |||
BC011192.1 | - | 5 | 403 | 4016679 | ||
BC085269.1 | - | 2 | 1558 | |||
BC090397.1 | - | 5 | 387 | 5150054 | ||
BC106137.1 | - | 5 | 388 | 4990874 | ||
BC125602.1 | - | 5 | 404 | 40130117 | ||
BC125604.1 | - | 5 | 404 | 40130119 |