Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042550.1 | NP_001036015.1 | 3 | 571 | ||
NM_001042551.1 | NP_001036016.1 | 3 | 509 | |||
NM_001265602.1 | NP_001252531.1 | 3 | 525 | |||
NM_006444.2 | NP_006435.2 | 3 | 500 | |||
XM_006716933.3 | XP_006716996.1 | 3 | 422 | |||
XM_011518148.2 | XP_011516450.1 | 3 | 385 | |||
XM_011518149.2 | XP_011516451.1 | 3 | 427 | |||
XM_011518152.2 | XP_011516454.1 | 3 | 451 | |||
XM_011518153.1 | XP_011516455.1 | 3 | 451 | |||
XM_017014206.1 | XP_016869695.1 | 3 | 571 | |||
XM_017014207.1 | XP_016869696.1 | 3 | 473 | |||
XM_017014208.1 | XP_016869697.1 | 3 | 513 | |||
GenBank mRNA | AF092563.1 | - | 3 | 435 | ||
AK001485.1 | - | 3 | 557 | |||
AK292599.1 | - | 3 | 448 | |||
AL833191.1 | - | 3 | 509 | |||
AY739716.1 | - | 3 | 401 | |||
BC003396.1 | - | 2 | 244 | |||
BC017845.1 | - | 3 | 439 | |||
BC032705.1 | - | 3 | 558 | |||
BC061906.1 | - | 3 | 457 | |||
BC071750.1 | - | 3 | 571 | |||
BC107891.1 | - | 3 | 545 | |||
BC130385.1 | - | 3 | 355 | 40146416 | ||
BC144163.1 | - | 3 | 371 | 9052683 |