Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170485.1 | NP_001163956.1 | 4 | 812 | ||
NM_001170486.1 | NP_001163957.1 | 5 | 924 | |||
NM_026552.3 | NP_080828.1 | 6 | 1043 | |||
GenBank mRNA | AJ278129.1 | - | 6 | 968 | ||
AK003923.1 | - | 1 | 126 | |||
AK017993.1 | - | 6 | 1046 | |||
AK018069.1 | - | 6 | 982 | |||
AK039215.1 | - | 6 | 992 | |||
AK076248.1 | - | 6 | 991 | |||
AK077325.1 | - | 6 | 966 | |||
AK133626.1 | - | 6 | 992 | |||
AK136082.1 | - | 6 | 987 | |||
AK150690.1 | - | 6 | 976 | |||
AK153380.1 | - | 6 | 975 | |||
AK159410.1 | - | 5 | 871 | |||
AK166440.1 | - | 6 | 963 | |||
BC015280.1 | - | 5 | 954 | |||
BC055309.1 | - | 5 | 954 | 6529255 |