Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apool | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026565.2 | NP_080841.1 | 8 | 651 | ||
GenBank mRNA | AK012398.1 | - | 8 | 697 | ||
AK012804.1 | - | 8 | 692 | |||
AK079141.1 | - | 8 | 703 | |||
BC024334.1 | - | 8 | 651 | 5036743 |