Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001992.1 | NP_001001992.1 | 3 | 324 | ||
NM_001032410.1 | NP_001027582.1 | 4 | 387 | |||
NM_006447.2 | NP_006438.1 | 3 | 324 | |||
XM_017028257.1 | XP_016883746.1 | 4 | 471 | |||
XM_017028258.1 | XP_016883747.1 | 4 | 353 | |||
XM_017028259.1 | XP_016883748.1 | 4 | 325 | |||
XM_017028260.1 | XP_016883749.1 | 2 | 78 | |||
XM_017028261.1 | XP_016883750.1 | 4 | 328 | |||
GenBank mRNA | AF126736.1 | - | 3 | 250 | ||
AK025104.1 | - | 3 | 290 | |||
AK025989.1 | - | 3 | 290 | |||
AK222681.1 | - | 3 | 253 | |||
AK222884.1 | - | 3 | 246 | |||
AK294284.1 | - | 2 | 188 | |||
AY333928.1 | - | 3 | 250 | |||
BC030777.2 | - | 3 | 280 | 4824914 | ||
CB960929.1 | - | 3 | 324 |