Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2e2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167921.1 | NP_001161393.1 | 2 | 160 | ||
NM_001167922.1 | NP_001161394.1 | 2 | 262 | |||
NM_026584.3 | NP_080860.3 | 2 | 148 | |||
GenBank mRNA | AK007492.1 | - | 1 | 105 | ||
AK021183.1 | - | 2 | 113 | |||
AK088342.1 | - | 2 | 173 | |||
AK146803.1 | - | 2 | 135 | |||
AK150415.1 | - | 2 | 106 | |||
AK153370.1 | - | 2 | 106 | |||
AK158783.1 | - | 2 | 242 | |||
BC020016.1 | - | 2 | 128 | 3501113 | ||
BX516160.1 | - | 2 | 399 | |||
BY256665.1 | - | 2 | 149 | |||
CX239577.1 | - | 2 | 160 |