Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cgnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018850.1 | - | 3 | 832 | ||
AK019857.1 | - | 3 | 819 | |||
AK173252.1 | - | 17 | 3056 | |||
BC031499.1 | - | 3 | 668 | |||
BC039211.1 | - | 11 | 1774 | |||
BC139006.1 | - | 18 | 4200 | 8862028 | ||
BC158061.1 | - | 18 | 4203 | 9007361 |