Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Leprotl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026609.2 | NP_080885.1 | 4 | 1470 | ||
GenBank mRNA | AK046807.1 | - | 1 | 932 | ||
AK150289.1 | - | 4 | 1529 | |||
AK153277.1 | - | 4 | 1515 | |||
AK154712.1 | - | 4 | 1535 | |||
AK159401.1 | - | 4 | 1457 | |||
BC004677.1 | - | 4 | 1470 | 3500223 |