Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026614.2 | NP_080890.1 | 3 | 314 | ||
GenBank mRNA | AK008155.1 | - | 3 | 185 | ||
AK008156.1 | - | 3 | 180 | |||
AK008286.1 | - | 3 | 180 | |||
AK008414.1 | - | 3 | 180 | |||
AK009338.1 | - | 3 | 198 | |||
AK009396.1 | - | 3 | 184 | |||
AK009461.1 | - | 3 | 184 | |||
AK009493.1 | - | 3 | 183 | |||
AK009608.1 | - | 3 | 184 | |||
AK009642.1 | - | 3 | 199 | |||
AK009652.1 | - | 3 | 184 | |||
AK009775.1 | - | 3 | 208 | |||
AK009840.1 | - | 3 | 198 | |||
AK009985.1 | - | 3 | 198 | |||
AK010024.1 | - | 3 | 183 | |||
AK010090.1 | - | 3 | 186 | |||
AK010092.1 | - | 3 | 200 | |||
AK011579.1 | - | 3 | 174 | |||
AK011947.1 | - | 3 | 174 | |||
AK019084.1 | - | 3 | 185 | |||
AK019201.1 | - | 3 | 185 | |||
AK019207.1 | - | 3 | 176 | |||
AK019215.1 | - | 3 | 184 | |||
AK019242.1 | - | 3 | 182 | |||
AK019245.1 | - | 3 | 205 | |||
AK019291.1 | - | 3 | 185 | |||
AK019301.1 | - | 3 | 315 | |||
AK019324.1 | - | 3 | 294 | |||
AK019354.1 | - | 3 | 187 | |||
AK019359.1 | - | 3 | 206 | |||
AK019364.1 | - | 3 | 200 | |||
AK019368.1 | - | 3 | 198 | |||
AK019430.1 | - | 3 | 196 | |||
BC028633.1 | - | 3 | 182 | 1247576 |