Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Toe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026654.2 | NP_080930.1 | 4, 5 | 369 | ||
GenBank mRNA | AK016889.1 | - | 359 | |||
AK019829.1 | - | 4, 5 | 369 | |||
AK046576.1 | - | 4, 5 | 400 | |||
AK082975.1 | - | 4, 5 | 350 | |||
BC023109.1 | - | 4, 5 | 345 | 4021435 |