Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Toe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026654.2 | NP_080930.1 | 8 | 1017 | ||
GenBank mRNA | AK019829.1 | - | 8 | 1017 | ||
AK046576.1 | - | 8 | 1048 | |||
AK082975.1 | - | 8 | 998 | |||
AK148609.1 | - | 3 | 370 | |||
BC023109.1 | - | 8 | 993 | 4021435 |