Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mto1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026658.2 | NP_080934.2 | 1 | 206 | ||
GenBank mRNA | AF369902.1 | - | 1 | 196 | ||
AK017573.1 | - | 1 | 204 | |||
AK019088.1 | - | 1 | 206 | |||
BC063256.1 | - | 1 | 192 | |||
BY735135.1 | - | 1 | 206 |