Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mto1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026658.2 | NP_080934.2 | 2 | 471 | ||
GenBank mRNA | AF369902.1 | - | 2 | 461 | ||
AK017573.1 | - | 2 | 469 | |||
AK019088.1 | - | 2 | 471 | |||
AK202743.1 | - | 1 | 82 | |||
BC063256.1 | - | 2 | 457 | |||
BY735135.1 | - | 2 | 471 |