Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam114a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026667.3 | NP_080943.3 | 14 | 2198 | ||
GenBank mRNA | AK030360.1 | - | 14 | 2199 | ||
AM162548.1 | - | 14 | 2202 | |||
BC027235.1 | - | 14 | 2161 | 3603083 | ||
BC110359.1 | - | 14 | 2199 | 5325099 |