Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163718.1 | NP_001157190.1 | 4 | 670 | ||
NM_026676.3 | NP_080952.2 | 4 | 691 | |||
GenBank mRNA | AK002309.1 | - | 4 | 663 | ||
AK003675.1 | - | 2 | 1142 | |||
AK158743.1 | - | 4 | 692 | |||
AK169072.1 | - | 3 | 1026 | |||
AK170074.1 | - | 4 | 660 | |||
BC006899.1 | - | 1 | 36 | |||
BC027751.1 | - | 4 | 761 | 5011894 | ||
BC038884.1 | - | 2 | 464 | 1516275 | ||
BC089019.1 | - | 4 | 640 | 30551363 | ||
CA979987.1 | - | 4 | 619 |