Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026697.3 | NP_080973.1 | 7, 8 | 834 | ||
GenBank mRNA | AB232609.1 | - | 6, 7 | 465 | ||
AJ968955.1 | - | 7, 8 | 689 | |||
AJ968956.1 | - | 6, 7 | 635 | |||
AJ968957.1 | - | 7, 8 | 689 | |||
AJ968958.1 | - | 6, 7 | 635 | |||
AJ968959.1 | - | 7, 8 | 689 | |||
AJ968960.1 | - | 6, 7 | 635 | |||
AK002704.1 | - | 7, 8 | 706 | |||
AK133963.1 | - | 7, 8 | 734 | |||
AK160859.1 | - | 7, 8 | 834 | |||
AK165812.1 | - | 7, 8 | 681 | |||
AK168551.1 | - | 7, 8 | 691 | |||
AK168867.1 | - | 7, 8 | 703 | |||
AK169396.1 | - | 7, 8 | 705 | |||
AK181684.1 | - | 4, 5 | 242 | |||
AK216803.1 | - | 4, 5 | 252 | |||
BC009085.1 | - | 7, 8 | 517 | 2649852 | ||
BC025139.1 | - | 7, 8 | 705 | 3980228 | ||
BC056648.1 | - | 7, 8 | 707 | 5034519 |