Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026697.3 | NP_080973.1 | 8 | 1701 | ||
GenBank mRNA | AK133963.1 | - | 8 | 1601 | ||
AK151834.1 | - | 1 | 735 | |||
AK160859.1 | - | 8 | 1701 | |||
AK165812.1 | - | 8 | 1548 | |||
AK166863.1 | - | 1 | 1720 | |||
AK168551.1 | - | 8 | 1558 | |||
AK168867.1 | - | 8 | 1570 | |||
AK169396.1 | - | 8 | 1572 | |||
BC009085.1 | - | 8 | 1384 | 2649852 |