Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291236.1 | NP_001278165.1 | 1 | 354 | ||
NM_026708.2 | NP_080984.1 | 1 | 354 | |||
GenBank mRNA | AK002703.1 | - | 1 | 250 | ||
AK046577.1 | - | 2 | 2234 | |||
AK161721.1 | - | 254 | ||||
AK165432.1 | - | 301 | ||||
BC005702.1 | - | 1 | 246 | 3598444 | ||
BC039182.1 | - | 1 | 324 | 3500908 |