Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ict1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026729.1 | NP_081005.1 | 3 | 163 | ||
GenBank mRNA | AK003192.1 | - | 3 | 163 | ||
AK003286.1 | - | 3 | 135 | |||
BC028523.1 | - | 3 | 264 | 2938122 |