Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026768.3 | NP_081044.1 | 6 | 646 | ||
GenBank mRNA | AB049953.1 | - | 6 | 655 | ||
AK003444.1 | - | 6 | 647 | |||
AK028226.1 | - | 6 | 648 | |||
AK172171.1 | - | 6 | 641 | |||
BC025456.1 | - | 4 | 442 | |||
BC058959.1 | - | 6 | 634 | 6817698 |