Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmvk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026784.3 | NP_081060.2 | 5 | 809 | ||
NM_027348.1 | NP_081624.1 | 5 | 778 | |||
GenBank mRNA | AK003607.1 | - | 5 | 612 | ||
AK013477.1 | - | 5 | 778 | |||
AK150483.1 | - | 5 | 728 | |||
AK168358.1 | - | 5 | 704 | |||
BC028659.1 | - | 4 | 597 | 1247653 |