Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026791.2 | NP_081067.2 | 7, 8 | 1154 | ||
GenBank mRNA | AK003747.1 | - | 2, 3 | 218 | ||
AK082085.1 | - | 6, 7 | 1643 | |||
AK156709.1 | - | 6, 7 | 1304 | |||
AK203100.1 | - | 1, 2 | 39 | |||
AK211947.1 | - | 1, 2 | 39 | |||
BC043658.1 | - | 7, 8 | 1115 | |||
BC053694.1 | - | 5, 6 | 570 | |||
BC060082.1 | - | 4, 5 | 385 | |||
BC144773.1 | - | 7, 8 | 1520 | |||
BC145661.1 | - | 7, 8 | 1520 | |||
CF197934.1 | - | 5, 6 | 558 |