Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myct1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026793.2 | NP_081069.1 | 2 | 848 | ||
GenBank mRNA | AK003845.1 | - | 2 | 802 | ||
AK030706.1 | - | 2 | 773 | |||
AK054222.1 | - | 2 | 747 | |||
AK142963.1 | - | 2 | 782 | |||
AY090114.1 | - | 2 | 801 | |||
BC096559.1 | - | 2 | 798 | 1399480 |