Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026836.2 | NP_081112.1 | 2 | 310 | ||
GenBank mRNA | AK050689.1 | - | 2 | 311 | ||
AK133851.1 | - | 3 | 1294 | |||
BC005603.1 | - | 2 | 289 | 3483315 |