Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srpx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083895.3 | NP_001077364.2 | 10 | 1683 | ||
NM_026838.4 | NP_081114.2 | 11 | 1735 | |||
GenBank mRNA | AF393640.1 | - | 9 | 1253 | ||
BC028307.1 | - | 11 | 1464 | 3488526 | ||
BQ560499.1 | - | 3 | 258 |