Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdgfrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026840.3 | NP_081116.3 | 6 | 1192 | ||
GenBank mRNA | AK004179.1 | - | 6 | 1084 | ||
AK132625.1 | - | 6 | 1193 | |||
BC039803.1 | - | 4 | 576 | |||
BC058275.1 | - | 6 | 1184 | 6515674 |