Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfand2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159905.1 | NP_001153377.1 | 4, 5 | 361 | ||
NM_001159906.1 | NP_001153378.1 | 4, 5 | 358 | |||
NM_026846.3 | NP_081122.2 | 3, 4 | 395 | |||
GenBank mRNA | AK004332.1 | - | 3, 4 | 385 | ||
AK151430.1 | - | 3, 4 | 397 | |||
BC011495.1 | - | 3, 4 | 420 | 4196340 | ||
CX217579.1 | - | 4, 5 | 364 |