Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026855.4 | NP_081131.1 | 2 | 228 | ||
GenBank mRNA | AK004398.1 | - | 2 | 229 | ||
BC119376.1 | - | 2 | 208 | 8734125 | ||
BC120631.1 | - | 2 | 208 | 40129554 | ||
BC144875.1 | - | 2 | 208 | |||
BC144876.1 | - | 2 | 208 |