Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291970.1 | NP_001278899.1 | 2, 3 | 441 | ||
NM_006502.2 | NP_006493.1 | 2, 3 | 441 | |||
XM_011514698.2 | XP_011513000.1 | 2, 3 | 433 | |||
GenBank mRNA | AB024313.1 | - | 2, 3 | 374 | ||
AF158185.1 | - | 2, 3 | 302 | |||
AK023893.1 | - | 2, 3 | 432 | |||
AK294430.1 | - | 2, 3 | 422 | |||
BC015742.1 | - | 2, 3 | 285 | 4872554 | ||
DC416114.1 | - | 2, 3 | 442 |