Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srrm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026886.3 | NP_081162.1 | 13 | 2706 | ||
GenBank mRNA | AK053836.1 | - | 13 | 2438 | ||
AK129457.1 | - | 5 | 1788 | |||
BC024130.1 | - | 2 | 1188 | |||
BC056392.1 | - | 13 | 2696 | 6855144 |