Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srrm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026886.3 | NP_081162.1 | 13 | 4602 | ||
GenBank mRNA | AK053836.1 | - | 14 | 4419 | ||
AK081496.1 | - | 1 | 548 | |||
AK129457.1 | - | 5 | 3684 |