Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcaf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026893.3 | NP_081169.2 | 5 | 1079 | ||
GenBank mRNA | AK005099.1 | - | 5 | 1025 | ||
AK030173.1 | - | 5 | 1031 | |||
AK031089.1 | - | 5 | 1009 | |||
AK078036.1 | - | 3 | 380 | |||
AK087517.1 | - | 5 | 963 | |||
AK090028.1 | - | 5 | 1028 | |||
AK143635.1 | - | 5 | 1033 | |||
AK173289.1 | - | 5 | 939 | |||
BC023852.1 | - | 5 | 1015 | 5346910 |