Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anapc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026904.2 | NP_081180.1 | 4 | 356 | ||
GenBank mRNA | AK005303.1 | - | 2 | 81 | ||
AK041373.1 | - | 408 | ||||
BC031460.1 | - | 3 | 356 | 3710036 |