Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anapc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026904.2 | NP_081180.1 | 5 | 1119 | ||
GenBank mRNA | AK041373.1 | - | 1171 | |||
BC031460.1 | - | 4 | 1119 | 3710036 |